Learn More
Grainger Engraving Tool 7.00mm L of Cut Carbide
Catalog No. 502764068
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R953 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch, an overall length of 3 inches, and a cutting diameter of 1/32 inch. This engraving tool is made of M2 high speed steel for wear resistance and has a bright finish. It can machine a variety of materials including steel, aluminum, brass, copper, plastics and wood. The single end cutting edge allows for detail work in cavities and confined areas. This is a high quality engraving tool well-suited for detailed machining in industrial and hobbyist applications.
Catalog No. 50-276-4068 Supplier Grainger Supplier No. 16R953
The Micro 100 Engraving Tool Single End 16R953 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch, an overall length of 3 inches, and a cutting diameter of 1/32 inch. This engraving tool is made of M2 high speed steel for wear resistance and has a bright finish. It can machine a variety of materials including steel, aluminum, brass, copper, plastics and wood. The single end cutting edge allows for detail work in cavities and confined areas. This is a high quality engraving tool well-suited for detailed machining in industrial and hobbyist applications.