Learn More
Grainger Micro 100 Engraving Tool, Carbide, 30°, 1 ea
Catalog No. 502764089
Encompass_Preferred
This carbide engraving tool is designed for precision engraving and finishing applications, primarily for use with steel materials. It features a conical, single-ended design with an uncoated finish for optimal performance.
  • Carbide material construction for durability
  • 30° included tip angle
  • 7 mm length of cut
  • 38 mm overall length
  • 3 mm shank diameter
  • Right hand cutting direction
  • Metric system of measurement
Catalog No. 50-276-4089 Supplier Grainger Supplier No. 16T018
This carbide engraving tool is designed for precision engraving and finishing applications, primarily for use with steel materials. It features a conical, single-ended design with an uncoated finish for optimal performance.
  • Carbide material construction for durability
  • 30° included tip angle
  • 7 mm length of cut
  • 38 mm overall length
  • 3 mm shank diameter
  • Right hand cutting direction
  • Metric system of measurement