Learn More
Grainger Engraving Tool 7/16 L of Cut Carbide
Catalog No. 502764066
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R921 is a carbide engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2", and a cutting diameter of 0.040". The tool is made of solid carbide and has a right hand spiral flute. It is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. The tool provides a smooth finish and can be used for detail engraving work in applications like sign making, plastic engraving, and metal stamping. With its small cutting diameter, this Micro 100 engraving tool allows for fine detail engraving.
Catalog No. 50-276-4066 Supplier Grainger Supplier No. 16R921
The Micro 100 Engraving Tool Single End 16R921 is a carbide engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2", and a cutting diameter of 0.040". The tool is made of solid carbide and has a right hand spiral flute. It is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. The tool provides a smooth finish and can be used for detail engraving work in applications like sign making, plastic engraving, and metal stamping. With its small cutting diameter, this Micro 100 engraving tool allows for fine detail engraving.