Learn More
Grainger Engraving Tool 7/16 L of Cut Carbide
Catalog No. 502763999
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R988 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 3 inches. The tool features a 135 degree point angle for detail engraving in materials like plastics, wood, aluminum and brass. It can be used in low to medium production runs at spindle speeds up to 15,000 RPM in CNC machines. The tool's high-speed steel construction provides wear resistance and heat resistance during engraving operations.
Catalog No. 50-276-3999 Supplier Grainger Supplier No. 16R988
The Micro 100 Engraving Tool Single End 16R988 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 3 inches. The tool features a 135 degree point angle for detail engraving in materials like plastics, wood, aluminum and brass. It can be used in low to medium production runs at spindle speeds up to 15,000 RPM in CNC machines. The tool's high-speed steel construction provides wear resistance and heat resistance during engraving operations.