Learn More
Grainger Engraving Tool 7/16 L of Cut Carbide
Catalog No. 502764005
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R985 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 3 inches. The tool features a 135 degree point angle for engraving a variety of materials including steel, aluminum, brass, copper, plastics, and wood. It can be used for hand engraving as well as in a milling machine or CNC machine. The tool has a bright finish for enhanced workpiece visibility.
Catalog No. 50-276-4005 Supplier Grainger Supplier No. 16R985
The Micro 100 Engraving Tool Single End 16R985 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 3 inches. The tool features a 135 degree point angle for engraving a variety of materials including steel, aluminum, brass, copper, plastics, and wood. It can be used for hand engraving as well as in a milling machine or CNC machine. The tool has a bright finish for enhanced workpiece visibility.