Learn More
MSC National Chemical Laboratories Floor Cleaner, Neutral, Concentrate, Lemon Scent, 1 Gallon
Catalog No. 502835974
Encompass
This alkaline-free cleaner is designed for maintaining wet-look gloss floor finishes. It features a low to moderate foaming, balanced detergent formula that emulsifies soils for easy, film-free cleaning. This product can be used through an automatic scrubber and provides a fresh lemon fragrance.
  • Suitable for healthcare settings.
  • Suitable for floors.
  • Restores shine and cleanliness without leaving streaks or residue.
  • Concentrate formula designed to be diluted, offering cost-effective and powerful cleaning.
  • Available in a 1-gallon package for long-lasting use.
  • Lemon scent.
  • Yellow in color.
  • Alkaline-free for gloss floor finishes.
  • Low to moderate foaming.
  • Emulsifies soils for easy, film-free cleaning.
  • May be used through an automatic scrubber.
Catalog No. 50-283-5974 Supplier MSC Supplier No. 63333298
This alkaline-free cleaner is designed for maintaining wet-look gloss floor finishes. It features a low to moderate foaming, balanced detergent formula that emulsifies soils for easy, film-free cleaning. This product can be used through an automatic scrubber and provides a fresh lemon fragrance.
  • Suitable for healthcare settings.
  • Suitable for floors.
  • Restores shine and cleanliness without leaving streaks or residue.
  • Concentrate formula designed to be diluted, offering cost-effective and powerful cleaning.
  • Available in a 1-gallon package for long-lasting use.
  • Lemon scent.
  • Yellow in color.
  • Alkaline-free for gloss floor finishes.
  • Low to moderate foaming.
  • Emulsifies soils for easy, film-free cleaning.
  • May be used through an automatic scrubber.