Learn More
Seracare Life Sciences Inc ENHANCE ORANGE BUFFER SOLUTION
Catalog No. 5067557
Encompass_Preferred
Enhance ORANGE Buffer Solution is a component of the HistoMark ORANGE immunohistochemical staining system for the detection of reagents labeled with horseradish peroxidase (HRP). This buffer must be used in place of 0.1 M Tris buffer when diluting KPL Diaminobenzidine-Catechol (DAB-C) Substrate Concentrate. Through enhancement with nickel, the normal light-brown reaction product of DAB is converted to a distinct orange stain
This item is not returnable. View return policy
Catalog No. 50-675-57 Supplier Seracare Life Sciences Inc Supplier No. 55100040
Enhance ORANGE Buffer Solution is a component of the HistoMark ORANGE immunohistochemical staining system for the detection of reagents labeled with horseradish peroxidase (HRP). This buffer must be used in place of 0.1 M Tris buffer when diluting KPL Diaminobenzidine-Catechol (DAB-C) Substrate Concentrate. Through enhancement with nickel, the normal light-brown reaction product of DAB is converted to a distinct orange stain