Learn More
Biogenex Laboratories Enhanced Antibody Diluent (Ready-to-Use)
SDP
Catalog No. NC0586413
Encompass_Preferred
BioGenex Enhanced Antibody Diluent is formulated for the dilution of all antibodies for immunohistochemistry. The diluent contains (1) chemical compounds to enhance antigen-antibody interaction and affinity, (2) preservative to enhance stability of the diluted antibodies during storage, and (3) carrier protein to minimize loss of the diluted antibody due to adsorption to the sides of dispensers and containers.BioGenex Enhanced Antibody Diluent is a ready-to-use solution. Dilutions are given as the ratio of concentrated volume to final volume (1:100 means 1 part of concentrated antibody mixed with 99 parts of diluent).
Catalog No. NC0586413 Supplier Biogenex Laboratories Supplier No. HK941YAK
BioGenex Enhanced Antibody Diluent is formulated for the dilution of all antibodies for immunohistochemistry. The diluent contains (1) chemical compounds to enhance antigen-antibody interaction and affinity, (2) preservative to enhance stability of the diluted antibodies during storage, and (3) carrier protein to minimize loss of the diluted antibody due to adsorption to the sides of dispensers and containers.BioGenex Enhanced Antibody Diluent is a ready-to-use solution. Dilutions are given as the ratio of concentrated volume to final volume (1:100 means 1 part of concentrated antibody mixed with 99 parts of diluent).