Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ Enhanced SPECI-GARD™ Biohazard Bags

Catalog No. 22310009
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-310-009 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-310-009 Supplier Minigrip™ Supplier No. SPECIUV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Transparent colored bags with biohazard logos

  • Separate document pocket with flap
  • Press and Close™ secure sample closure provides infection prevention and user safety
  • Utilizes “no glove to adhesive” contact capability when sealing bags
  • Easy-to-open tear line
  • Simple instructions for use printed on bags

Order Info

All bags measure 6W x 10 in. H.

Specifications

Color White
For Use With (Application) Easy-to-open tear line
Includes Press and Close liquid tight adhesive closure, perforated opening, 2 pockets, biohazard label
Material Polyethylene
Width (English) 6 in.
Height (English) 10 in.
Height (Metric) 25.4 cm
Product Type Biohazard Bag
Width (Metric) 15.24 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.