Learn More
MSC 3M DBI-SALA ExoFit NEX Safety Harness, Climbing, Positioning, 1 ea
Catalog No. 5000688851
Encompass
The 3M™ DBI-SALA® ExoFit™ NEX™ Safety Harness features a lightweight design for comfort and durability during demanding work environments. This harness is engineered for applications including climbing, positioning, rescue, and arc-flash protection, providing essential fall safety for industrial and occupational use.
  • Revolving torso adapters allow for easy and rapid fit adjustment
  • Automatic stand-up back D-ring facilitates efficient dorsal connections
  • Integrated suspension trauma straps included
  • Lightweight aluminum D-rings offer strength without added bulk
  • Available in a wide range of sizes from X-Small to 3X
Catalog No. 50-006-88851 Supplier MSC Supplier No. 23135213
The 3M™ DBI-SALA® ExoFit™ NEX™ Safety Harness features a lightweight design for comfort and durability during demanding work environments. This harness is engineered for applications including climbing, positioning, rescue, and arc-flash protection, providing essential fall safety for industrial and occupational use.
  • Revolving torso adapters allow for easy and rapid fit adjustment
  • Automatic stand-up back D-ring facilitates efficient dorsal connections
  • Integrated suspension trauma straps included
  • Lightweight aluminum D-rings offer strength without added bulk
  • Available in a wide range of sizes from X-Small to 3X