Learn More
AdipoGen EnhancedTRAIL Soluble h Pack
SDP
Catalog No. 502562753
Encompass_Preferred
Set. Induces apoptosis in a concentration range of 5-50ng/ml in the presence of 1µg/ml of TNF Ligands Enhancer Prod. No. AG-35B-0001.
Catalog No. 50-256-2753 Supplier AdipoGen Supplier No. AG44B0002KI01
Set. Induces apoptosis in a concentration range of 5-50ng/ml in the presence of 1µg/ml of TNF Ligands Enhancer Prod. No. AG-35B-0001.