Learn More
Enzo Life Sciences Enhancer for Ligands (50 µg)
SDP
Supplier:  Enzo Life Sciences ALX804034C050
Encompass_Preferred
Enhances biological activity of TRAIL, soluble (human) (recombinant) and FasL, soluble (human) (recombinant). Formulation: Liquid. In PBS, pH 7.4, containing 50% glycerol. Contains no preservatives. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. NC0467993