Learn More
Cayman Chemical Enniatin Complex, Purity: ≥95%, CAS: 11113-62-5, 50 mg
SDP
Catalog No. 502894803
Encompass_Preferred
A mixture of cyclohexadepsipeptides commonly isolated from fungi that act as ionophores, forming pores in cellular membranes to allow selective ion transport; exhibits antimicrobial activity against a variety of eukaryotic and prokaryotic genera, inhibits acyl-CoA:cholesterol acyltranferase, and induces apoptosis in several cancer lines
Catalog No. 50-289-4803 Supplier Cayman Chemical Supplier No. 900204050 MG
A mixture of cyclohexadepsipeptides commonly isolated from fungi that act as ionophores, forming pores in cellular membranes to allow selective ion transport; exhibits antimicrobial activity against a variety of eukaryotic and prokaryotic genera, inhibits acyl-CoA:cholesterol acyltranferase, and induces apoptosis in several cancer lines