Learn More
Medchemexpress LLC HY-10533 10mg Medchemexpress, Eniluracil CAS:59989-18-3 Purity:>98%
SDP
Catalog No. 501932263
Encompass_Preferred
Medchemexpress, HY-10533 10mg Eniluracil CAS:59989-18-3 Eniluracil (5-Ethynyluracil), a uracil analogue and a mechanism-based irreversible inhibitor of dihydropyrimidine dehydrogenase (DPD) , increases the oral bioavailability of 5-fluorouracil (5-FU) to 100%, facilitating uniform absorption and predictable toxicity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2263 Supplier Medchemexpress LLC Supplier No. HY1053310MG
Medchemexpress, HY-10533 10mg Eniluracil CAS:59989-18-3 Eniluracil (5-Ethynyluracil), a uracil analogue and a mechanism-based irreversible inhibitor of dihydropyrimidine dehydrogenase (DPD) , increases the oral bioavailability of 5-fluorouracil (5-FU) to 100%, facilitating uniform absorption and predictable toxicity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.