Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Eniporide hydrochloride | 211813-86-4 | MFCD31560490 | 99.5% | 356.83 g/mol | C14H17ClN4O3S | 1 ML
SDP

Catalog No. 5000242726
Encompass_Preferred

Eniporide hydrochloride is a potent Na+/H+ (NHE) exchange inhibitor supplied for research use. It is commonly used in cellular and biochemical studies of NHE activity and cardiovascular research, and is offered as both solid material and as a ready-to-use DMSO solution.

  • Potent Na+/H+ exchange inhibitor for NHE studies.
  • Available as a 10 mM solution in DMSO or as a solid.
  • Approximately 99.5% purity suitable for analytical and biological assays.
  • Molecular weight 356.83 g/mol; formula C14H17ClN4O3S.
  • CAS number 211813-86-4 for unambiguous identification.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-42726 Supplier Medchemexpress LLC Supplier No. HY106150B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eniporide hydrochloride is a potent Na+/H+ (NHE) exchange inhibitor supplied for research use. It is commonly used in cellular and biochemical studies of NHE activity and cardiovascular research, and is offered as both solid material and as a ready-to-use DMSO solution.

  • Potent Na+/H+ exchange inhibitor for NHE studies.
  • Available as a 10 mM solution in DMSO or as a solid.
  • Approximately 99.5% purity suitable for analytical and biological assays.
  • Molecular weight 356.83 g/mol; formula C14H17ClN4O3S.
  • CAS number 211813-86-4 for unambiguous identification.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.