Learn More
Medchemexpress LLC HY-103019 2mg , (+)-Enitociclib CAS:1610358-56-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1030192MG
Encompass_Preferred
HY-103019 2mg (+)-Enitociclib CAS: 1610358-56-9 (+)-Enitociclib ((+)-BAY-1251152) is an enanthiomer of BAY-1251152 with rotation (+). (+)-Enitociclib is a potent and selective CDK9 inhibitor with an IC50 of 3 nM. (+)-Enitociclib has anti-tumour activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9300