Learn More
TARGETMOL CHEMICALS INC ENMD-1068 HCl 50MG
SDP
Catalog No. 502877517
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. ENMD-1068 HCl (ENMD 1068) is a PAR-2 antagonist. ENMD-1068 HCl inhibits the development of endometriosis in a mouse model.
Catalog No. 50-287-7517 Supplier TARGETMOL CHEMICALS INC Supplier No. T2726750MG
Also available in 1 mL, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. ENMD-1068 HCl (ENMD 1068) is a PAR-2 antagonist. ENMD-1068 HCl inhibits the development of endometriosis in a mouse model.