Learn More
Medchemexpress LLC ENMD-1068 hydrochloride | 2703451-51-6 | 98.3% | 319.87 | C15H30ClN3O2 | 10MM 1ML
SDP
Catalog No. 5000236847
Encompass_Preferred
ENMD-1068 hydrochloride is a selective protease-activated receptor 2 (PAR2) antagonist used in biochemical and cellular research to probe PAR2 signaling and related pathways such as TGF-β1/Smad. The compound is supplied as the hydrochloride salt with high reported purity and is offered both as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid pack sizes for flexibility in experimental workflows.
  • Selective PAR2 antagonist suitable for signaling and fibrosis research.
  • Provided as the hydrochloride salt with white to off-white appearance.
  • Available as a 10 mM DMSO solution (1 mL) for immediate use in assays.
  • Offered in solid quantities for custom formulation and dosing.
  • High reported purity (~98.3%) and defined molecular weight for reliable dosing.
  • Solubility data and storage recommendations provided for in vitro and in vivo use.
Catalog No. 50-002-36847 Supplier Medchemexpress LLC Supplier No. HY124748A10MM 1ML
ENMD-1068 hydrochloride is a selective protease-activated receptor 2 (PAR2) antagonist used in biochemical and cellular research to probe PAR2 signaling and related pathways such as TGF-β1/Smad. The compound is supplied as the hydrochloride salt with high reported purity and is offered both as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid pack sizes for flexibility in experimental workflows.
  • Selective PAR2 antagonist suitable for signaling and fibrosis research.
  • Provided as the hydrochloride salt with white to off-white appearance.
  • Available as a 10 mM DMSO solution (1 mL) for immediate use in assays.
  • Offered in solid quantities for custom formulation and dosing.
  • High reported purity (~98.3%) and defined molecular weight for reliable dosing.
  • Solubility data and storage recommendations provided for in vitro and in vivo use.