Learn More
Abcam ENMD-1068, protease-activated receptor 2 (PAR2) anatagonist, 25MG
Catalog No. 502706476
Encompass_Preferred
MW 283.41 Da, Purity >99%. Selective protease-activated receptor 2 (PAR2) antagonist (IC₅₀ = 1.2 mM). Anti-inflamatory agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6476 Supplier Abcam Supplier No. AB14169925MG
MW 283.41 Da, Purity >99%. Selective protease-activated receptor 2 (PAR2) antagonist (IC₅₀ = 1.2 mM). Anti-inflamatory agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement