Learn More
Medchemexpress LLC ENMD-1198 | 864668-87-1 | 98.9% | 311.42 | 100 MG
SDP
Catalog No. 5000392596
Encompass_Preferred
ENMD-1198 is an orally active microtubule destabilizing agent and a 2-methoxyestradiol analogue with antiproliferative and antiangiogenic activity. It is suitable for inhibiting HIF-1alpha and STAT3 in human HCC cells and leads to reduced tumor growth and vascularization.
  • Inhibits HIF-1alpha and STAT3.
  • Reduces tumor growth.
  • Reduces vascularization.
  • Exhibits antiproliferative activity against human MDA-MB-231 cells.
  • Inhibits HCC cell growth with IC50s of 2.5 μM for HUH-7 and HepG2 cells.
Catalog No. 50-003-92596 Supplier Medchemexpress LLC Supplier No. HY16196100MG
ENMD-1198 is an orally active microtubule destabilizing agent and a 2-methoxyestradiol analogue with antiproliferative and antiangiogenic activity. It is suitable for inhibiting HIF-1alpha and STAT3 in human HCC cells and leads to reduced tumor growth and vascularization.
  • Inhibits HIF-1alpha and STAT3.
  • Reduces tumor growth.
  • Reduces vascularization.
  • Exhibits antiproliferative activity against human MDA-MB-231 cells.
  • Inhibits HCC cell growth with IC50s of 2.5 μM for HUH-7 and HepG2 cells.