Learn More
Medchemexpress LLC ENMD-1198 (IRC-110160) | 864668-87-1 | 98.87% | 311.42 | 5 MG
SDP
Catalog No. 5000392888
Encompass_Preferred
ENMD-1198 (IRC-110160) is an orally active microtubule destabilizing agent, which is a 2-methoxyestradiol analogue with antiproliferative and antiangiogenic activity. It is effective in inhibiting HIF-1alpha and STAT3 in human HCC cells, leading to reduced tumor growth and vascularization.
  • Orally active microtubule destabilizing agent.
  • 2-methoxyestradiol analogue.
  • Antiproliferative activity.
  • Antiangiogenic activity.
  • Inhibits HIF-1alpha and STAT3 in human HCC cells.
  • Reduces tumor growth and vascularization.
  • Inhibits HCC cell growth.
  • Abrogates EGF-induced phosphorylation of Akt, FAK, p44/42 MAPK, and STAT3.
  • Effectively inhibits growth of hepatocellular carcinoma and angiogenesis in vivo.
Catalog No. 50-003-92888 Supplier Medchemexpress LLC Supplier No. HY161965MG
ENMD-1198 (IRC-110160) is an orally active microtubule destabilizing agent, which is a 2-methoxyestradiol analogue with antiproliferative and antiangiogenic activity. It is effective in inhibiting HIF-1alpha and STAT3 in human HCC cells, leading to reduced tumor growth and vascularization.
  • Orally active microtubule destabilizing agent.
  • 2-methoxyestradiol analogue.
  • Antiproliferative activity.
  • Antiangiogenic activity.
  • Inhibits HIF-1alpha and STAT3 in human HCC cells.
  • Reduces tumor growth and vascularization.
  • Inhibits HCC cell growth.
  • Abrogates EGF-induced phosphorylation of Akt, FAK, p44/42 MAPK, and STAT3.
  • Effectively inhibits growth of hepatocellular carcinoma and angiogenesis in vivo.