Learn More
TARGETMOL CHEMICALS INC ENMD-2076 5MG
SDP
Catalog No. 502876281
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ENMD-2076, a multi-targeted kinase inhibitor, has specific activity against Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRalpha. Purity 98%
Catalog No. 50-287-6281 Supplier TARGETMOL CHEMICALS INC Supplier No. T23585MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ENMD-2076, a multi-targeted kinase inhibitor, has specific activity against Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRalpha. Purity 98%