Learn More
Selleck Chemical LLC ENMD-2076 S1181-5mg
SDP
Catalog No. 503111445
Encompass_Preferred
ENMD-2076 has selective activity against Aurora A and Flt3 with IC50 of 14 nM and 1 86 nM 25-fold selective for Aurora A than over Aurora B and less potent to RET SRC NTRK1/TRKA CSF1R/FMS VEGFR2/KDR FGFR and PDGFR ENMD-2076 inhibits the growth of a wide range of human solid tumor and hematopoietic cancer cell lines with IC50 from 0 025 to 0 7 M which induces apoptosis and G2/M phase arrest Phase 2
Catalog No. 50-311-1445 Supplier Selleck Chemical LLC Supplier No. S11815MG
ENMD-2076 has selective activity against Aurora A and Flt3 with IC50 of 14 nM and 1 86 nM 25-fold selective for Aurora A than over Aurora B and less potent to RET SRC NTRK1/TRKA CSF1R/FMS VEGFR2/KDR FGFR and PDGFR ENMD-2076 inhibits the growth of a wide range of human solid tumor and hematopoietic cancer cell lines with IC50 from 0 025 to 0 7 M which induces apoptosis and G2/M phase arrest Phase 2