Learn More
Apexbio Technology LLC ENMD-2076 934353-76-1 50mg
SDP
Catalog No. 5000093139
Encompass_Preferred
ENMD-2076 (CAS 934353-76-1) is a selective inhibitor of Aurora A kinase (IC50 14 nM) and FLT3 (IC50 1 86 nM) with additional inhibitory activity against multiple tyrosine kinases implicated in angiogenesis and cell proliferation including RET FLT4/VEGFR3 SRC CSF1R NTRK1 FGFR1/2 VEGFR2/KDR LCK and PDGFR (IC50 1 86 120 nM) ENMD-2076 disrupts cell cycle progression by arresting cells in the G2/M phase through inhibition of Aurora kinases suppresses the PI3K/AKT signaling pathway and demonstrates cytotoxic effects in hematologic malignancy and solid tumor cell lines (in vitro IC50 25 700 nM) In vivo it reduces tumor growth and angiogenesis in xenograft models supporting its use in cancer research and antiangiogenic studies
Catalog No. 50-000-93139 Supplier Apexbio Technology LLC Supplier No. A413050
ENMD-2076 (CAS 934353-76-1) is a selective inhibitor of Aurora A kinase (IC50 14 nM) and FLT3 (IC50 1 86 nM) with additional inhibitory activity against multiple tyrosine kinases implicated in angiogenesis and cell proliferation including RET FLT4/VEGFR3 SRC CSF1R NTRK1 FGFR1/2 VEGFR2/KDR LCK and PDGFR (IC50 1 86 120 nM) ENMD-2076 disrupts cell cycle progression by arresting cells in the G2/M phase through inhibition of Aurora kinases suppresses the PI3K/AKT signaling pathway and demonstrates cytotoxic effects in hematologic malignancy and solid tumor cell lines (in vitro IC50 25 700 nM) In vivo it reduces tumor growth and angiogenesis in xenograft models supporting its use in cancer research and antiangiogenic studies