Learn More
Medchemexpress LLC ENMD-2076 | 934353-76-1 | 99.99% | 375.47 | 10 MG
SDP
Catalog No. 5000345134
Encompass_Preferred
ENMD-2076 is a multi-targeted kinase inhibitor, demonstrating significant activity against Aurora A, Flt3, KDR/VEGFR2, and other kinases. It effectively inhibits cellular autophosphorylation and induces G2-M-phase arrest and apoptosis in various cell lines. In vivo studies show dose-dependent inhibition of tumor growth and tumor regression, with the compound being well tolerated.
  • Inhibits multiple kinases including Aurora A, Flt3, VEGFR2
  • Induces G2-M-phase arrest and apoptosis
  • Demonstrates antiproliferative activity against human cell lines
  • Inhibits cellular Flt3 ligand (FL)-induced Flt3 autophosphorylation
  • Inhibits stem cell factor (SCF)-induced Kit autophosphorylation
  • Inhibits VEGFR2/KDR autophosphorylation
  • Leads to dose-dependent inhibition of tumor growth in vivo
Catalog No. 50-003-45134 Supplier Medchemexpress LLC Supplier No. HY10987A50MG
ENMD-2076 is a multi-targeted kinase inhibitor, demonstrating significant activity against Aurora A, Flt3, KDR/VEGFR2, and other kinases. It effectively inhibits cellular autophosphorylation and induces G2-M-phase arrest and apoptosis in various cell lines. In vivo studies show dose-dependent inhibition of tumor growth and tumor regression, with the compound being well tolerated.
  • Inhibits multiple kinases including Aurora A, Flt3, VEGFR2
  • Induces G2-M-phase arrest and apoptosis
  • Demonstrates antiproliferative activity against human cell lines
  • Inhibits cellular Flt3 ligand (FL)-induced Flt3 autophosphorylation
  • Inhibits stem cell factor (SCF)-induced Kit autophosphorylation
  • Inhibits VEGFR2/KDR autophosphorylation
  • Leads to dose-dependent inhibition of tumor growth in vivo