Learn More
Medchemexpress LLC ENMD-2076 | 934353-76-1 | 99.99% | 375.47 | 100 MG
SDP
Catalog No. 5000344576
Encompass_Preferred
ENMD-2076 is a multi-targeted kinase inhibitor with high selectivity towards Aurora A versus Aurora B. It inhibits HUVEC growth and shows antiproliferative effects on human leukemia cell lines. This compound induces G2-M-phase arrest and apoptosis in U937 cells and inhibits Flt3 and Kit autophosphorylation. In vivo, ENMD-2076 demonstrates dose-dependent inhibition of tumor growth and is well tolerated in animal studies.
  • Multi-targeted kinase inhibitor.
  • Inhibits HUVEC growth.
  • Shows antiproliferative effects on leukemia cell lines.
  • Induces G2-M-phase arrest and apoptosis.
  • Inhibits Flt3 ligand (FL)-induced Flt3 autophosphorylation.
  • Inhibits stem cell factor (SCF)-induced Kit autophosphorylation.
  • Inhibits VEGFR2/KDR autophosphorylation.
  • Results in dose-dependent inhibition of tumor growth in vivo.
Catalog No. 50-003-44576 Supplier Medchemexpress LLC Supplier No. HY10987A1MG
ENMD-2076 is a multi-targeted kinase inhibitor with high selectivity towards Aurora A versus Aurora B. It inhibits HUVEC growth and shows antiproliferative effects on human leukemia cell lines. This compound induces G2-M-phase arrest and apoptosis in U937 cells and inhibits Flt3 and Kit autophosphorylation. In vivo, ENMD-2076 demonstrates dose-dependent inhibition of tumor growth and is well tolerated in animal studies.
  • Multi-targeted kinase inhibitor.
  • Inhibits HUVEC growth.
  • Shows antiproliferative effects on leukemia cell lines.
  • Induces G2-M-phase arrest and apoptosis.
  • Inhibits Flt3 ligand (FL)-induced Flt3 autophosphorylation.
  • Inhibits stem cell factor (SCF)-induced Kit autophosphorylation.
  • Inhibits VEGFR2/KDR autophosphorylation.
  • Results in dose-dependent inhibition of tumor growth in vivo.