Learn More
Medchemexpress LLC ENMD-2076 | 934353-76-1 | 99.99% | 375.47 | 50 MG
SDP
Catalog No. 5000344537
Encompass_Preferred
ENMD-2076 is a multi-targeted kinase inhibitor with IC50s of 1.86, 14, 58.2, 15.9, 92.7, 70.8, 56.4 nM for Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRα, respectively. It is for research use only and not sold to patients.
  • Multi-targeted kinase inhibitor
  • Orally active
  • Antiangiogenic and antiproliferative mechanisms of action
Catalog No. 50-003-44537 Supplier Medchemexpress LLC Supplier No. HY10987A10MM/1ML
ENMD-2076 is a multi-targeted kinase inhibitor with IC50s of 1.86, 14, 58.2, 15.9, 92.7, 70.8, 56.4 nM for Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRα, respectively. It is for research use only and not sold to patients.
  • Multi-targeted kinase inhibitor
  • Orally active
  • Antiangiogenic and antiproliferative mechanisms of action