Learn More
Selleck Chemical LLC ENMD-2076 L-()-Tartaric acid S2018-5mg
SDP
Catalog No. 503108413
Encompass_Preferred
ENMD-2076 L-( )-Tartaric acid is the tartaric acid of ENMD-2076 selective activity against Aurora A and Flt3 with IC50 of 14 nM and 1 86 nM 25-fold more selective for Aurora A than Aurora B and less potent to VEGFR2/KDR and VEGFR3 FGFR1 and FGFR2 and PDGFR Phase 2
Catalog No. 50-310-8413 Supplier Selleck Chemical LLC Supplier No. S20185MG
ENMD-2076 L-( )-Tartaric acid is the tartaric acid of ENMD-2076 selective activity against Aurora A and Flt3 with IC50 of 14 nM and 1 86 nM 25-fold more selective for Aurora A than Aurora B and less potent to VEGFR2/KDR and VEGFR3 FGFR1 and FGFR2 and PDGFR Phase 2