Learn More
Medchemexpress LLC ENMD-2076 Tartrate | 1453868-32-0 | 99.66% | 525.56 | 50 MG
SDP
Catalog No. 5000344768
Encompass_Preferred
ENMD-2076 Tartrate is a multi-targeted kinase inhibitor primarily targeting Aurora A, Flt3, and VEGFR2. It is intended for research use only and not for patient applications. This compound is known for its inhibitory effects on various kinases, contributing to its potential in anti-proliferative and anti-angiogenic research.
  • Exhibits potent inhibition of Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, and PDGFRα.
  • Induces G2-M-phase arrest and apoptosis in U937 cells.
  • Inhibits Flt3 and Kit autophosphorylation.
  • Shows dose-dependent inhibition of tumor growth in vivo.
Catalog No. 50-003-44768 Supplier Medchemexpress LLC Supplier No. HY1098710MG
ENMD-2076 Tartrate is a multi-targeted kinase inhibitor primarily targeting Aurora A, Flt3, and VEGFR2. It is intended for research use only and not for patient applications. This compound is known for its inhibitory effects on various kinases, contributing to its potential in anti-proliferative and anti-angiogenic research.
  • Exhibits potent inhibition of Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, and PDGFRα.
  • Induces G2-M-phase arrest and apoptosis in U937 cells.
  • Inhibits Flt3 and Kit autophosphorylation.
  • Shows dose-dependent inhibition of tumor growth in vivo.