Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ENMD-2076 Tartrate | 1453868-32-0 | 99.7% | 525.56 | 10 MG
SDP

Catalog No. 5000344573
Encompass_Preferred

ENMD-2076 Tartrate is a multi-targeted kinase inhibitor with IC50s of 1.86, 14, 58.2, 15.9, 92.7, 70.8, 56.4 nM for Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRα, respectively. It is for research use only and not sold to patients.

  • Acts as a multi-targeted kinase inhibitor
  • Inhibits Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, and PDGFRα
  • Shows selective inhibition of Aurora A over Aurora B
  • Inhibits human umbilical vein endothelial cell (HUVEC) growth
  • Blocks cellular Flt3 ligand (FL)-induced Flt3 autophosphorylation
  • Prevents stem cell factor (SCF)-induced Kit autophosphorylation
  • Inhibits VEGFR2/KDR autophosphorylation
  • Demonstrates dose-dependent inhibition of tumor growth or tumor regression in vivo
  • Exhibits good tolerance in daily doses during in vivo studies

Catalog No. 50-003-44573 Supplier Medchemexpress LLC Supplier No. HY1098710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ENMD-2076 Tartrate is a multi-targeted kinase inhibitor with IC50s of 1.86, 14, 58.2, 15.9, 92.7, 70.8, 56.4 nM for Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRα, respectively. It is for research use only and not sold to patients.

  • Acts as a multi-targeted kinase inhibitor
  • Inhibits Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, and PDGFRα
  • Shows selective inhibition of Aurora A over Aurora B
  • Inhibits human umbilical vein endothelial cell (HUVEC) growth
  • Blocks cellular Flt3 ligand (FL)-induced Flt3 autophosphorylation
  • Prevents stem cell factor (SCF)-induced Kit autophosphorylation
  • Inhibits VEGFR2/KDR autophosphorylation
  • Demonstrates dose-dependent inhibition of tumor growth or tumor regression in vivo
  • Exhibits good tolerance in daily doses during in vivo studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.