Learn More
Medchemexpress LLC HY-10987 5mg Medchemexpress, ENMD-2076 (Tartrate) CAS:1291074-87-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY109875MG
Encompass_Preferred
Medchemexpress, HY-10987 5mg ENMD-2076 (Tartrate) CAS:1291074-87-7 ENMD-2076 Tartrate is a multi-targeted kinase inhibitor with IC50s of 1.86, 14, 58.2, 15.9, 92.7, 70.8, 56.4 nM for Aurora A, Flt3, KDR/VEGFR2, Flt4/VEGFR3, FGFR1, FGFR2, Src, PDGFRα, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8725