Learn More
Cayman Chemical Enmetazobactam, Purity: ≥95%, CAS: 1001404-83-6, 10 mg
SDP
Catalog No. 502990185
Encompass_Preferred
A β-lactamase inhibitor; has greater activity against class A β-lactamases (IC50s = 0.006-0.52 µM) than class B or class D β-lactamases (IC50s = 8.7->100 µM); active against 223 Enterobacteriaceae clinical isolates in combination with cefepime (MIC50 = 0.125 mg/L); reduces mortality in mouse models of septicemia induced by E. coli, K. pneumoniae, or E. cloacae in combination with cefepime
Catalog No. 50-299-0185 Supplier Cayman Chemical Supplier No. 3071510 MG
A β-lactamase inhibitor; has greater activity against class A β-lactamases (IC50s = 0.006-0.52 µM) than class B or class D β-lactamases (IC50s = 8.7->100 µM); active against 223 Enterobacteriaceae clinical isolates in combination with cefepime (MIC50 = 0.125 mg/L); reduces mortality in mouse models of septicemia induced by E. coli, K. pneumoniae, or E. cloacae in combination with cefepime