Learn More
Enzo Life Sciences Enniatin B (1mg). CAS: 917-13-5
SDP
Supplier:  Enzo Life Sciences ALX380007M001
Encompass_Preferred
Ionophore antibiotic. Inhibitor of the S. cerevisiae ABC transporter Pdr5p.Inhibitor of acyl-CoA-cholesterol acyltransferase and of phosphodiesterase. Potential anticancer compound. Purity: ≥90% (HPLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in DMSO or other organic solvents; almost insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8901