Learn More
AdipoGen Enniatin B
SDP
Catalog No. 502562514
Encompass_Preferred
Chemical. CAS 917-13-5. Formula C33H57N3O9. MW 639.8. Isolated from fungus Fusarium sp. Cyclohexadepsipeptide mycotoxin. One of four major analogs in the enniatin complex. Commonly found food contaminant in cereals and their products. Ionophore antibiotic less ionophoric than Enniatin A. Incorporation into the cell membrane forms dimeric structures that transport monovalent ions across the membrane especially the mitochondrial membranes affecting oxidative phosphorylation uncoupling. Anticancer compound. Triggers apoptosis in several cancer cell lines. Moderate inhibitor of ACAT acylcoenzyme Acholesterolacyl transferase. Inhibitor of the pleiotropic drug resistance protein 5 Pdr5p in yeast. Shown to have a variety of other biological activities such as antifungal, anthelmintic, insecticidal, immunomodulatory and phytotoxic activity.
Catalog No. 50-256-2514 Supplier AdipoGen Supplier No. AGCN20479M005
Chemical. CAS 917-13-5. Formula C33H57N3O9. MW 639.8. Isolated from fungus Fusarium sp. Cyclohexadepsipeptide mycotoxin. One of four major analogs in the enniatin complex. Commonly found food contaminant in cereals and their products. Ionophore antibiotic less ionophoric than Enniatin A. Incorporation into the cell membrane forms dimeric structures that transport monovalent ions across the membrane especially the mitochondrial membranes affecting oxidative phosphorylation uncoupling. Anticancer compound. Triggers apoptosis in several cancer cell lines. Moderate inhibitor of ACAT acylcoenzyme Acholesterolacyl transferase. Inhibitor of the pleiotropic drug resistance protein 5 Pdr5p in yeast. Shown to have a variety of other biological activities such as antifungal, anthelmintic, insecticidal, immunomodulatory and phytotoxic activity.