Learn More
Aviva Systems Biology Corp Eno1 Antibody - N-terminal region (ARP38030_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38030_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Eno1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Eno1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-708