Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2026MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: ENO2/1462. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b. Positive Control: HepG2, SH-SY-5Y, HeLa or Y79 cells. Pancreas, Cerebellum or Pheochromocytoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide corresponding to aa416-433 of human NSE gamma. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 50kDa.Chromosome: 12p13. Gene Symbol: ENO2. Gene ID: 2026. Ensemble: ENSG00000111674. Uniprot: NP_001966.
Catalog No. 50-188-4670