Learn More
Aviva Systems Biology Corp ENO2 Antibody - middle region (ARP60777_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60777_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ENO2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ENO2. Alternate names: NSEApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-158