Learn More
Medchemexpress LLC HY-123523 5mg Medchemexpress, Enocitabine CAS:55726-47-1 Purity:>98%
SDP
Catalog No. 501969646
Encompass_Preferred
Medchemexpress, HY-123523 5mg Enocitabine CAS:55726-47-1 Enocitabine is a nucleoside analog, and is a potent DNA replication inhibitor, and a DNA chain terminator. Enocitabine inhibits the replication of human cytomegalovirus. Enocitabine has antileukemic and antiviral activities[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9646 Supplier Medchemexpress LLC Supplier No. HY1235235MG
Medchemexpress, HY-123523 5mg Enocitabine CAS:55726-47-1 Enocitabine is a nucleoside analog, and is a potent DNA replication inhibitor, and a DNA chain terminator. Enocitabine inhibits the replication of human cytomegalovirus. Enocitabine has antileukemic and antiviral activities[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.