Learn More
Cayman Chemical Enocitabine, Purity: ≥95%, CAS: 55726-47-1, 10 mg
SDP
Catalog No. 502911663
Encompass_Preferred
A prodrug form of cytarabine; resistant to deamination by cytidine deaminase; reduces the virus yield in the supernatant of CMV-infected Vero cells (EC90 = 0.5 µg/ml); inhibits viral DNA synthesis at 5 µg/ml; increases survival in an L1210 murine skin lymphocytic leukemia model
Catalog No. 50-291-1663 Supplier Cayman Chemical Supplier No. 3653810 MG
A prodrug form of cytarabine; resistant to deamination by cytidine deaminase; reduces the virus yield in the supernatant of CMV-infected Vero cells (EC90 = 0.5 µg/ml); inhibits viral DNA synthesis at 5 µg/ml; increases survival in an L1210 murine skin lymphocytic leukemia model