Learn More
Cayman Chemical Enoctabn 50mg
SDP
Catalog No. 502911660
Encompass_Preferred
A prodrug form of cytarabine; resistant to deamination by cytidine deaminase; reduces the virus yield in the supernatant of CMV-infected Vero cells (EC90 = 0.5 UG/ml); inhibits viral DNA synthesis at 5 UG/ml; increases survival in an L1210 murine skin lymphocytic leukemia model
Catalog No. 50-291-1660 Supplier Cayman Chemical Supplier No. 3653850 MG
A prodrug form of cytarabine; resistant to deamination by cytidine deaminase; reduces the virus yield in the supernatant of CMV-infected Vero cells (EC90 = 0.5 UG/ml); inhibits viral DNA synthesis at 5 UG/ml; increases survival in an L1210 murine skin lymphocytic leukemia model