Learn More
TRIBIOSCIENCE INC Enolase Activity Assay (Colorimetric / Fluorometric), 100 Tests
SDP
Catalog No. NC3454578
Encompass_Preferred
Tribioscience’s Enolase Activity Assay is designed to be a flexible, accurate, simple, and time-saving method in which enolase catalyzes 2-PG into PEP, resulting in an intermediate formation through a coupled enzyme reaction. It is catalyzed and reacts with the probe, to generate a colorimetric (570 nm) or fluorometric (Ex/Em=530/590 nm) intensity proportional to the enolase activity which can be measured. The kit is enough for 100 assays.
Catalog No. NC3454578 Supplier TRIBIOSCIENCE INC Supplier No. TBS2092100
Tribioscience’s Enolase Activity Assay is designed to be a flexible, accurate, simple, and time-saving method in which enolase catalyzes 2-PG into PEP, resulting in an intermediate formation through a coupled enzyme reaction. It is catalyzed and reacts with the probe, to generate a colorimetric (570 nm) or fluorometric (Ex/Em=530/590 nm) intensity proportional to the enolase activity which can be measured. The kit is enough for 100 assays.