Learn More
Cayman Chemical EnopeptIn A 2.5mg
SDP
Catalog No. 502979572
Encompass_Preferred
A depsipeptide antibiotic that contains two unusual amino acids (N-methylalanine and 4-methylproline) and features a pentaenone side chain; effective against Gram-positive bacteria, including methicillin-resistant S. aureus (MIC = 25 UG/ml), and Gram-negative bacteria, including mutant forms of E. coli and P. aeruginosa (MICs = 200 UG/ml) yet does not inhibit fungi
Catalog No. 50-297-9572 Supplier Cayman Chemical Supplier No. 170922.5 MG
A depsipeptide antibiotic that contains two unusual amino acids (N-methylalanine and 4-methylproline) and features a pentaenone side chain; effective against Gram-positive bacteria, including methicillin-resistant S. aureus (MIC = 25 UG/ml), and Gram-negative bacteria, including mutant forms of E. coli and P. aeruginosa (MICs = 200 UG/ml) yet does not inhibit fungi