Learn More
Aviva Systems Biology Corp ENOPH1 Antibody - middle region (ARP57530_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57530_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ENOPH1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ENOPH1. Alternate names: DKFZp586M0524, E1, FLJ12594, MASA, MST145ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-286-329