Learn More
Enzo Life Sciences eNOS (human) (recombinant) (10µg)
SDP
Catalog No. 502008042
Encompass_Preferred
Alternative name: Nitric oxide synthase (endothelial), NOS III. Purity: ≥90% (SDS-PAGE, Western Blot). MW: ~135kDa/subunit; homodimer. Formulation: Liquid. In 50 mM TRIS, pH 7.4, containing 0.1mM EDTA, 10% glycerol, 100mM NaCl, 0.1mM DTT, 10 µM tetrahydrobiopterin. Source: Produced in Sf9 insect cells. Untagged human eNOS. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-200-8042 Supplier Enzo Life Sciences Supplier No. ALX2018530010
Alternative name: Nitric oxide synthase (endothelial), NOS III. Purity: ≥90% (SDS-PAGE, Western Blot). MW: ~135kDa/subunit; homodimer. Formulation: Liquid. In 50 mM TRIS, pH 7.4, containing 0.1mM EDTA, 10% glycerol, 100mM NaCl, 0.1mM DTT, 10 µM tetrahydrobiopterin. Source: Produced in Sf9 insect cells. Untagged human eNOS. Produced in a baculovirus expression system. Long Term Storage: -80°C.