Learn More
Aviva Systems Biology Corp ENOX1 Antibody - middle region (ARP40886_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40886_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ENOX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ENOX1. Alternate names: CNOX, FLJ10094, PIG38, bA64J21.1, cCNOXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-57652