Learn More
Aviva Systems Biology Corp ENOX2 Antibody - N-terminal region (ARP59871_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59871_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ENOX2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ENOX2. Alternate names: APK1, COVA1, tNOXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-860