Learn More
AdipoGen Enoxacin
SDP
Supplier:  AdipoGen AGCR13539M250
Encompass_Preferred
Chemical. CAS 74011-58-8. Formula C15H17FN4O3. MW 320.3. Enoxacin is a cell permeable fluoroquinolone broad-spectrum antibacteral agent. It is active against clinical isolates of a variety of Gram-positive and Gram-negative bacteria. Fluoroquinolones are a group of broad-spectrum antibiotics that inhibit DNA synthesis. They prevent unwinding of double-stranded DNA by binding DNA gyrase or DNA topoisomerase IV, prevent the formation of single-stranded DNA and inhibit bacterial replication. Enoxacin is used to treat urinary tract infections and gonorrhea. Osteoclastogenesis inhibitor. Shown to decrease RANKL-induced JNK signaling and inhibiting osteoclast formation. Additionally, Enoxacin inhibits vacuolar H ATPase activity, preventing bone resorption. Enoxacin is a unique small molecule enhancer of microRNA SMER maturation. MicroRNAs miRNAs are small RNA molecules that regulate gene expression at the posttranscriptional level and are critical for many cellular pathways.
Catalog No. 50-256-2322