Learn More
Medchemexpress LLC Enoxacin-d8 hydrochloride | 2930288-95-0 | 99.98% | 1 MG
SDP
Catalog No. 5000405831
Encompass_Preferred
Enoxacin-d8 (hydrochloride) is a deuterium-labeled Enoxacin. Enoxacin is a fluoroquinolone that interferes with DNA replication and inhibits bacterial DNA gyrase and topoisomerase IV. It also acts as a miRNA processing activator, enhancing siRNA-mediated mRNA degradation and promoting the biogenesis of endogenous miRNAs. This compound exhibits potent activity against both gram-positive and gram-negative bacteria and is identified as a cancer-specific growth inhibitor by enhancing TAR RNA-binding protein 2 (TRBP)-mediated microRNA processing. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05831 Supplier Medchemexpress LLC Supplier No. HYB0268S11MG
Enoxacin-d8 (hydrochloride) is a deuterium-labeled Enoxacin. Enoxacin is a fluoroquinolone that interferes with DNA replication and inhibits bacterial DNA gyrase and topoisomerase IV. It also acts as a miRNA processing activator, enhancing siRNA-mediated mRNA degradation and promoting the biogenesis of endogenous miRNAs. This compound exhibits potent activity against both gram-positive and gram-negative bacteria and is identified as a cancer-specific growth inhibitor by enhancing TAR RNA-binding protein 2 (TRBP)-mediated microRNA processing. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS