Learn More
Cayman Chemical Enoxacin-d8 (hydrochloride), Purity: ≥99% deuterated forms (d1-d8), CAS: 2930288-95-0, 10 mg
SDP
Catalog No. 502950683
Encompass_Preferred
An internal standard for the quantification of enoxacin by GC- or LC-MS
Catalog No. 50-295-0683 Supplier Cayman Chemical Supplier No. 3354410 MG
An internal standard for the quantification of enoxacin by GC- or LC-MS