Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enoxacin, G-Biosciences
SCI_ED

Catalog No. S139783
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139783 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139783 Supplier G-Biosciences Supplier No. RC1002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enoxacin is a fluoroquinolone antibiotic compound that is markedly effective against gram negative bacteria.

It has been used to study fluoroquinolone cross-resistence in Escehria coli, Pseudomonas aeruginosa and Staphylococcus aureus. Enoxacin is structurally related to nalidixic acid and inhibits DNA gyrase in bacteria.

Chemical Identifiers

Molecular Weight (g/mol) 347.36

Specifications

Physical Form Powder
Quantity 1 g
Product Line Gibco
Boiling Point 481°C
Percent Purity >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.